missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Picric Acid, Reagent Grade, Ricca Chemical
For safety in transportation, Picric Acid is usually mixed with a minimum of 30% water.
$324.10 - $324.10
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C6H3N3O7 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | OXNIZHLAWKMVMX-UHFFFAOYSA-N |
| IUPAC Name | 2,4,6-trinitrophenol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RDCP030010C
|
Ricca Chemical Company
RDCP030010C1 |
10 g |
Each for $324.10
|
|
|||||
Specifications
| 7732-18-5,88-89-1 | |
| C6H3N3O7 | |
| OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O | |
| Mixture | |
| Reagent | |
| Amber Glass Bottle |
| 70 | |
| OXNIZHLAWKMVMX-UHFFFAOYSA-N | |
| 2,4,6-trinitrophenol | |
| 49% | |
| 10 g | |
| Picric Acid |
Safety and Handling
ShelfLife : 84 months