missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid-EDTA Reagent, Reagents

Catalog No. 01334513
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334513 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334513 Supplier Labchem Inc Supplier No. CP0065004BP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Picric Acid-EDTA Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 6381-92-6 , 1303-96-4
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 2,4,6-Trinitrophenol
IUPAC Name water
SMILES O

Specifications

pH 8.7 to 10.5
Boiling Point 101°C
Color Yellow
Quantity 4 L
Packaging Opaque HDPE
Physical Form Liquid
Specific Gravity 1.0143
Grade Reagent
Chemical Name or Material Picric Acid-EDTA Reagent