missing translation for 'onlineSavingsMsg'
Learn More

Picramic Acid (wetted with ca. 25% Water) (unit weight on dry weight basis) 98.0+%, TCI America™

Supplier:  TCI America D081325G

Catalog No. D081325G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Picramic Acid (wetted with ca. 25% Water) (unit weight on dry weight basis)
173°C
C6H5N3O5
25 g
2-Amino-4,6-dinitrophenol
C1=C(C=C(C(=C1[N+](=O)[O-])O)N)[N+](=O)[O-]
4921319
≥98.0% (T)
96-91-3
Red-Yellow
MFCD00070534
3317
QXYMVUZOGFVPGH-UHFFFAOYSA-N
199.122
199.12
Crystalline Powder

Chemical Identifiers

96-91-3
199.122
QXYMVUZOGFVPGH-UHFFFAOYSA-N
4921319
C6H5N3O5
MFCD00070534
2-Amino-4,6-dinitrophenol
C1=C(C=C(C(=C1[N+](=O)[O-])O)N)[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0820

RTECSNumber : SJ5800000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions