accessibility menu, dialog, popup

UserName

CAS RN 96-91-3

IUPAC Name: 2-amino-4,6-dinitrophenol
Synonyms: picramic acid
Molecular Weight (g/mol): 199.12
Molecular Formula: C6H5N3O5
InChi Key: QXYMVUZOGFVPGH-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1O)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results