missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PI Lipid Kinase Substrate

Catalog No. PV5371
Encompass
Change view
Click to view available options
Quantity:
800 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5371 800 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5371 Supplier Thermo Scientific™ Supplier No. PV5371

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifically optimized for use with the Class II (PIK3C2A & PIK3C2B) PI3 kinases.

Specifically optimized for use with the Class II (PIK3C2A & PIK3C2B) PI3 kinases. These vesicles consist of 100% phosphatidylinositol (PI).

For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

Specifications

Assay Entry Biochemical ADP accumulation
Chromagen Quantity 800nmol
Content And Storage PI Lipid Kinase Substrate in 50 mM HEPES, pH 7.5

Store at -20°C.
Detection Method Fluorescence
Form Frozen
For Use With (Application) Pharma and biopharma, target and lead identification and validation, drug discovery and development, kinase biology, biochemical kinase assays, Adapta™ universal kinase assays
Quantity 800 nmol
Substrate Phosphatidylinositol (PI) vesicles
Substrate Properties Lipid-Based Substrate, Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Lipid Kinases
Product Type PI Lipid Kinase Substrate
Shipping Condition Dry Ice
Storage Requirements –20°C
Target Entry PIK3C2B, PIK3C2A
Validated Kinases PIK3C2B
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.