missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion™ Plus DNA Polymerase

Catalog No. F630XL
Change view
Click to view available options
No. of Reactions:
100 Reactions
2000 Reactions
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
F630XL 2000 Reactions
F630L 500 Reactions
F630S 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F630XL Supplier Thermo Scientific™ Supplier No. F630XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing.

Thermo Scientific Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing. It is a Pyrococcus-like proofreading DNA polymerase fused to a processivity-enhancing domain, enabling Phusion Plus DNA Polymerase to generate PCR sequences with high accuracy, sensitivity, and inhibitor tolerance.

Features include:

  • >100X higher sequence accuracy than Taq polymerase
  • Universal annealing temperature of 60°C in PCR
  • Hot-start mechanism reduces non-specific amplification and primer degradation
  • Automation enabled due to 24-hour stability of assembled reactions
  • High inhibitor tolerance for work with DNA of suboptimal purity

The novel Phusion Plus reaction buffer includes proprietary additives that stabilize primer/template duplexes during the annealing step at 60°C. This unique buffer formulation increases PCR specificity without the need to optimize annealing temperature for each primer pair, helping save time and avoid mistakes in PCR runs. Furthermore, with Phusion Plus DNA Polymerase and its universal annealing feature, different PCR reactions can be cycled together using the same protocol.

Phusion Plus DNA Polymerase is inactive at ambient temperatures due to its hot-start mechanism mediated by Affibody molecules, allowing setup and storage of assembled PCR reactions at room temperature. Enzyme activity is fully restored after the initial denaturation step.

Applications

  • High-fidelity PCR
  • Cloning & mutagenesis
  • Template generation for sequencing
  • GC-rich PCR
  • Long-range PCR (up to 20 kb)
  • High-throughput PCR

Notes

  • To simplify reactrion setup, Phusion Plus DNA Polymerase is also available in ready-to-use master mix formats.

Specifications

Content And Storage Four packs, each containing:
• Phusion Plus DNA Polymerase (250 μL)
• 5X Phusion Plus Buffer (5 x 1.25 mL)
• 5X Phusion GC Enhancer (4 x 1.25 mL)

Store at –15°C to –25°C.
GC-Rich PCR Performance High
Polymerase Phusion Plus DNA Polymerase
Reaction Speed 15 to 30 sec./kb
Quantity 2000 x 50 μL reactions
Shipping Condition Room Temp or Wet Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) >100X
Hot Start Built-In Hot Start
No. of Reactions 2000 Reactions
Overhang Blunt
Reaction Format Separate components
Size (Final Product) 20 kb or less
Show More Show Less
What is the fidelity of the Phusion Plus DNA Polymerase (Cat. No. F630S)?

Please see "Conclusions" within the technical note for the Phusion Plus DNA Polymerase (Cat. No. F630S) (see link: https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/measuring-error-rates-ngs-dna-polymerases-tech-note.pdf).

Can I set up my reactions on ice if I am using the Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix (Cat. No. F631S)?

Reaction setup with Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix can be done either on ice or at room temperature.

For Research Use Only. Not for use in diagnostic procedures.