missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Human Specimen Direct PCR Kit

Catalog No. F150
Change view
Click to view available options
Quantity:
200 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F150 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F150 Supplier Thermo Scientific™ Supplier No. F150BID

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Phusion Human Specimen Direct PCR Kit delivers unprecedented convenience for DNA amplification by allowing PCR directly from unpurified human samples.

Thermo Scientific Phusion Human Specimen Direct PCR Kit delivers unprecedented convenience for DNA amplification by allowing PCR directly from unpurified human samples. A tiny amount of sample is used directly in the PCR reaction with no prior purification steps. This allows significant savings in both time and cost.

Tested sample materials include buccal swabs, saliva, amniotic fluid, hair, fingernails, teeth, and skin biopsies. The kit is compatible with fresh, frozen, and even formalin-fixed, paraffin-embedded (FFPE) human tissue samples. The Phusion Human Specimen Direct PCR Kit includes optimized reagents and detailed protocols for many sample materials. The modified Phusion Hot Start II High-Fidelity DNA Polymerase utilized in the kit is a highly accurate proofreading DNA polymerase, and PCR products obtained are perfectly suitable even for cloning purposes.

Features

  • No need for time-consuming and expensive DNA purification steps
  • Two simple protocols for various applications
  • Extremely short PCR protocol times
  • Modified Phusion Hot Start II High-Fidelity DNA Polymerase delivers abundant yields of specific product

Notes

  • The optimal annealing temperature for Phusion DNA Polymerases may differ significantly from that of Taq-based DNA polymerases.

Specifications

Content And Storage • Phusion Human Specimen DNA Polymerase, 80 μL
• 2X Phusion Human Specimen PCR Buffer, 2 x 1.25 mL
• Universal Control Primer Mix (25 μM), 40 μL
• Dilution Buffer, 2 x 5 mL
• DNARelease Additive, 3 x 100 μL

Store at -20°C.
GC-Rich PCR Performance High
PCR Method Direct PCR
Polymerase Phusion Human Specimen DNA Polymerase
Reaction Speed Fast
Product Type Direct PCR Kit
Quantity 200 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Hot-start PCR
Hot Start Built-In Hot Start
No. of Reactions 200 reactions
Overhang Blunt
Reaction Format Separate Components
Sample Type Human
Starting Material DNA
Show More Show Less
What is your recommendation to take a sample from plant or animal tissue for Direct PCR?

In order to obtain best results from Direct PCR, it is important to use a very small amount of starting materials. The recommended size is 0.3-0.5 mm in diameter. We recommend using Tissue Puncher 0.3 mm (Cat. No. F-200S), which is a convenient tool for sampling and is suitable for both plant and animal tissues.

Why are Control primers provided in the Direct PCR Kits?

It is very important to include a control reaction in the Direct PCR to ensure that the PCR conditions are optimal for the specific tissue or plant material used. Control primers are universal primers that amplify a small fragment from a conserved genome region. If the reaction with control primers fails, the sample material may not be suitable for Direct PCR, or PCR conditions need optimization.

How are samples, stored at different conditions, compatible with the Phire Tissue Direct PCR Master Mix?

With the Phire Tissue Direct PCR Master Mix you can amplify DNA from differently stored tissues (fresh, frozen, FFPE) from human and a variety of animal species.

Can I use plant or tissue sample which is not on the list of the Direct PCR Master Mix and Kit tested samples?

You can test any plant or tissue sample. However, it is recommended to first optimize the reaction. When working with new sample materials, start with the Dilution & Storage protocol as it allows several PCR reactions to be performed from the same sample during optimization. It is recommended to perform a positive control with purified sample DNA to ensure that the PCR conditions are working. If the positive control with purified DNA fails, the PCR conditions should be optimized before continuing further.

Can you amplify the same sample materials with the Direct PCR Master Mixes as with Direct PCR Kits?

Yes, the Direct PCR Master Mixes are upgrades of the Direct PCR kits and thus are suitable for the same sample materials.

Do the Direct PCR Master Mixes and Kits come with tissue punchers?

No, tissue punchers are not provided. You can purchase 0.3 mm Tissue Punchers (Cat. No. F-200S), or cut the sample with a scalpel. Some scientists use 100 µL pipette tips to take a sample.

Do the Direct PCR Master Mixes work in Dilution & Storage protocol?

Yes, similar to the Direct PCR Kits, Direct PCR Master Mixes work in both Direct and Dilution & Storage protocols. Direct PCR from blood is performed with Direct protocol only.

Which FFPE sample types are suitable for Direct PCR from FFPE tissues?

FFPE tissue, FFPE tissue sections, tissue blocks and microscope slides can all be used in sample preparation.

What if I want to clone or sequence the PCR product from a Direct PCR kit? Can I use the kit with Phusion instead of Phire?

Direct PCR Kits and Master Mixes employ Phire Hot Start II DNA Polymerase or Phusion Hot Start II Polymerase (depending on the kit), that both possess the following activities: 5'-3' DNA polymerase activity and a weak 3'-5' exonuclease activity. When cloning fragments amplified with these DNA Polymerases, blunt end cloning is recommended. If TA cloning is required, it can be performed by adding A overhangs to the blunt PCR product. However, before adding the overhangs, it is very important to remove all the DNA polymerase by purifying the PCR product carefully. Any remaining Phusion or Phire DNA Polymerases will degrade the A overhangs, creating blunt ends again. All Direct PCR kits are compatible with sequencing techniques. The green dye present in the Direct Master Mix kits does not interfere with the sequencing reactions.

After performing Direct PCR, how should I store the PCR products amplified directly from the sample?

After PCR, it is recommended to centrifuge the PCR reactions at 1000 x g for 1-3 minutes to collect the supernatant for subsequent analysis. We recommend storing the supernatant at -20 degrees C

Are there any restrictions to the size of the amplicon in Direct PCR?

The optimal length for Direct PCR from liquid samples is up to 5 kb and from solid samples up to 1 kb. However, the maximum length depends on the sample type, sample size and the reaction volume. For example we have amplified a 7.5 kb fragment directly from human blood and tissue samples, and a 3 kb fragment from mouse ear and 5 kb fragment from plant leaves.

When performing Direct PCR, when do I need to use DNARelease Additive?

DNARelease Additive is required when PCR is performed directly from certain tissue samples using the Direct protocol. Cell debris present in these PCR products can cause DNA fragments to get trapped in the agarose gel wells. DNARelease Additive eliminates this problem and should be added directly into the PCR reaction. Please reduce the amount of water used or simply add the DNARelease to the PCR reaction setup. DNARelease is also used in the Dilution & Storage protocol to improve the release of DNA from the tissue sample.


For Research Use Only. Not for use in diagnostic procedures.