missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion High-Fidelity PCR Master Mix with HF or GC Buffer

Catalog No. p-9026017
Change view
Click to view available options
Includes:
GC Buffer
HF Buffer
No. of Reactions:
100 Reactions
500 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes No. of Reactions
F532S GC Buffer 100 Reactions
F532L GC Buffer 500 Reactions
F531L HF Buffer 500 Reactions
F531S HF Buffer 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. F532S Supplier Thermo Scientific™ Supplier No. F532S
Only null left

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

Phusion High-Fidelity PCR Master Mix is convenient 2X mix containing Phusion DNA Polymerase, nucleotides, and optimized reaction buffer including MgCl2. Two master mix formulations are available: with HF Buffer (F-531S and F-531L) and with GC Buffer (F-532S and F-532L). The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the master mix with HF Buffer should be used as the default master mix for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures.

Highlights

  • High fidelity (25X Taq)
  • Fast PCR due to short extension times (15–30 s/kb)
  • Robust performance, minimal optimization needed

Applications

  • Amplification of difficult (GC-rich) templates
  • High-fidelity PCR
  • Cloning
  • Template generation for sequencing
  • Long-range PCR (up to 20 kb)
  • Mutagenesis
  • High throughput PCR

Using Phusion DNA polymerases

Annealing rules for Phusion DNA polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

Specifications

Concentration 2X
Content And Storage • 2X Phusion Master Mix with GC Buffer (2 x 1.25 mL)
• 100% DMSO (1 x 500 μL)

Store at –20°C.
GC-Rich PCR Performance High
Polymerase Phusion High-Fidelity DNA Polymerase
Reaction Speed Fast
Product Type High-Fidelity PCR Master Mix
Quantity 100 reactions
Shipping Condition Dry Ice
For Use With (Application) Standard PCR, High-fidelity PCR
Fidelity (vs. Taq) 52X
Hot Start No
No. of Reactions 100 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 20 kb or less
Includes GC Buffer
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.