missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Blood Direct PCR Kit

Thermo Scientific Phusion Blood Direct PCR Kit is developed for amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR.

Supplier:  Thermo Scientific™ F547L

 View more versions of this product

Catalog No. F547L


Only null left

Description

Description

Thermo Scientific Phusion Blood Direct PCR Kit is for use in the amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR. The modified Phusion Hot Start II High-Fidelity DNA Polymerase exhibits high resistance to PCR inhibitors found in blood and retains polymerase activity even with samples containing 40% whole blood.

This DNA polymerase exhibits exceptional accuracy in proofreading and makes the Phusion Blood Direct PCR Kit one of the best choices for demanding applications, such as cloning. The Phusion Blood Direct PCR Kit is versatile and permits analysis of blood samples from various species and with different anticoagulants or storage conditions.

Features

  • No need for time-consuming and expensive DNA purification steps
  • Simple protocol requires minimal hands-on time
  • Extremely short PCR protocol times
  • Modified Phusion Hot Start II High-Fidelity DNA Polymerase delivers abundant yields of specific product

Notes

  • The optimal annealing temperature for Phusion DNA Polymerases may differ significantly from that of Taq-based polymerases.
Specifications

Chemical Identifiers

2 U/μL
Direct PCR
Fast
Wet or Dry Ice
Blunt
Blood
High
Phusion Blood II DNA Polymerase
Direct PCR Kit
Built-In Hot Start
Separate Components
DNA

Specifications

2 U/μL
High
Phusion Blood II DNA Polymerase
Direct PCR Kit
Wet or Dry Ice
500 Reactions
Separate Components
DNA
• Phusion Blood II DNA Polymerase, 200 μL
• 2X Phusion Blood PCR Buffer with dNTPs and MgCl2, 5 x 1.25 mL
• Universal Control Primer Mix, 40 μL
• 50 mM EDTA, 0.5 mL
• 50 mM MgCl2, 1.5 mL
• DMSO, 0.5 mL

Store at -20°C.
Direct PCR
Fast
500 reactions
Built-In Hot Start
Blunt
Blood
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.