missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalylsulfathiazole 95.0+%, TCI America™
$74.56 - $344.86
Chemical Identifiers
| CAS | 85-73-4 |
|---|---|
| Molecular Formula | C17H13N3O5S2 |
| Molecular Weight (g/mol) | 403.427 |
| MDL Number | MFCD00005318 |
| InChI Key | PBMSWVPMRUJMPE-UHFFFAOYSA-N |
| PubChem CID | 4806 |
| ChEBI | CHEBI:9336 |
| IUPAC Name | 2-[[4-(1,3-thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid |
| SMILES | C1=CC=C(C(=C1)C(=O)NC2=CC=C(C=C2)S(=O)(=O)NC3=NC=CS3)C(=O)O |
Chemical Identifiers
| 85-73-4 | |
| 403.427 | |
| PBMSWVPMRUJMPE-UHFFFAOYSA-N | |
| CHEBI:9336 | |
| C1=CC=C(C(=C1)C(=O)NC2=CC=C(C=C2)S(=O)(=O)NC3=NC=CS3)C(=O)O |
| C17H13N3O5S2 | |
| MFCD00005318 | |
| 4806 | |
| 2-[[4-(1,3-thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid |
Specifications
| 85-73-4 | |
| White | |
| MFCD00005318 | |
| 4′C-(2-Thiazolylsulfamoyl)phthalanilic Acid | |
| C1=CC=C(C(=C1)C(=O)NC2=CC=C(C=C2)S(=O)(=O)NC3=NC=CS3)C(=O)O | |
| 403.427 | |
| CHEBI:9336 | |
| ≥95.0% (T) | |
| Phthalylsulfathiazole |
| 240°C | |
| C17H13N3O5S2 | |
| 25 g | |
| PBMSWVPMRUJMPE-UHFFFAOYSA-N | |
| 2-[[4-(1,3-thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid | |
| 4806 | |
| 403.43 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TH8575000
TSCA : Yes