missing translation for 'onlineSavingsMsg'
Learn More

Phthalylsulfathiazole 95.0+%, TCI America™

Catalog No. T070325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T070325G 25 g
T0703500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T070325G Supplier TCI America Supplier No. T070325G

Chemical Identifiers

CAS 85-73-4
Molecular Formula C17H13N3O5S2
Molecular Weight (g/mol) 403.427
MDL Number MFCD00005318
InChI Key PBMSWVPMRUJMPE-UHFFFAOYSA-N
PubChem CID 4806
ChEBI CHEBI:9336
IUPAC Name 2-[[4-(1,3-thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)NC2=CC=C(C=C2)S(=O)(=O)NC3=NC=CS3)C(=O)O

Specifications

Melting Point 240°C
Color White
Quantity 25 g
Synonym 4′C-(2-Thiazolylsulfamoyl)phthalanilic Acid
Formula Weight 403.43
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalylsulfathiazole
RTECSNumber TH8575000
TSCA Yes