missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Pentoxide - Methanesulfonic Acid, TCI America™

Catalog No. P104225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P104225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P104225G Supplier TCI America Supplier No. P104225G
Only null left

Chemical Identifiers

CAS 39394-84-8
Molecular Formula CH4O8P2S
Molecular Weight (g/mol) 238.043
MDL Number MFCD00070545
InChI Key JHNLZOVBAQWGQU-UHFFFAOYSA-N
Synonym eaton's reagent, phosphorus pentoxide methanesulfonic acid, diphosphooxidane; methanesulfonic acid, eaton's, eaton'sreagent, eaton inverted exclamation markas reagent, eaton's reagent, p2o5 ca w/w, phosphorus pentoxide, 7.7 wt. % in methanesulfonic acid
PubChem CID 16149098
SMILES CS(=O)(=O)O.O=P(=O)OP(=O)=O

Specifications

Color Undesignated
Boiling Point 122°C
UN Number 2922
Quantity 25 g
Physical Form Clear Liquid at 20°C
Chemical Name or Material Phosphorus Pentoxide - Methanesulfonic Acid