accessibility menu, dialog, popup

UserName

CH3SOOHOOPOOPOO

CAS RN 39394-84-8

IUPAC Name: diphosphooxidane; methanesulfonic acid
Synonyms: diphosphooxidane; methanesulfonic acid
Molecular Weight (g/mol): 238.04
Molecular Formula: CH4O8P2S
InChi Key: JHNLZOVBAQWGQU-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.O=P(=O)OP(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)