missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus ICP Standard, 10,000 ppm P in H2O, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 10,000ppm P |
| CAS Min % | 94.0, 3.51 |
| CAS Max % | 98.0, 3.65 |
| Equivalence | 1 mL = 10 mg P |
| Physical Form | Liquid |
| Color | Colorless |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 50 mL |
| Show More |