missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus AA Standard, 1000 ppm P in H2O, Ricca Chemical
Supplier: Ricca Chemical Company AP1KW100
Specifications
| Phosphorus AA Standard | |
| 7722-76-1 , 7732-18-5 | |
| 100.0,0.38 | |
| H6NO4P | |
| Colorless | |
| Standard | |
| N.OP(O)(O)=O | |
| 115.03 | |
| 24402 | |
| 1mL = 1mg P (1, 000ppm P)NH4H2PO4 in H2O |
| Natural LDPE Bottle | |
| 98.0,0.36 | |
| Liquid | |
| MFCD00003396 | |
| 100°C | |
| LFVGISIMTYGQHF-UHFFFAOYSA-N | |
| phosphoric acid amine | |
| 100 mL | |
| CHEBI:62982 |
Chemical Identifiers
| 7722-76-1 | |
| 115.03 | |
| LFVGISIMTYGQHF-UHFFFAOYSA-N | |
| CHEBI:62982 | |
| N.OP(O)(O)=O |
| H6NO4P | |
| MFCD00003396 | |
| 24402 | |
| phosphoric acid amine |
Safety and Handling
ShelfLife : 24 months