missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 75%, Technical, Approx. 75%, Spectrum™ Chemical

$310.83 - $16953.27

Chemical Identifiers

CAS 7664-38-2
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18-612-332
View Documents
Spectrum Chemical Mfg Cor
P1481500MLGL
500 mL Amber Glass Bottle
Each for $310.83
Only null left
 
18-612-329
View Documents
Spectrum Chemical Mfg Cor
P148125LTPV
2.5 L Poly-Coated Glass Bottle
Each for $656.03
Only null left
 
18-612-333
View Documents
Spectrum Chemical Mfg Cor
P14814LT
4 L Poly-Coated Glass Bottle
Each for $851.07
Only null left
 
18-612-330
View Documents
Spectrum Chemical Mfg Cor
P148120LT
20 L Poly Drum
Each for $2,317.50
Only null left
 
18-612-331
View Documents
Spectrum Chemical Mfg Cor
P148155GLBL
55 gal. Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7664-38-2

Specifications

7664-38-2,7732-18-5
Amber Glass Bottle
∼75%
Phosphoric Acid
1
500 mL
Technical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.