missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 75%, Technical, Approx. 75%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P148125LTPV
Specifications
| Phosphoric Acid | |
| 1 | |
| 2.5 L | |
| Technical |
| 7664-38-2,7732-18-5 | |
| Poly-Coated Glass Bottle | |
| ∼75% |
Chemical Identifiers
| 7664-38-2 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.