missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 75%, Technical, Approx. 75%, Spectrum™ Chemical

Catalog No. 18612332
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612332 500 mL Amber Glass Bottle
18612329 2.5 L Poly-Coated Glass Bottle
18612333 4 L Poly-Coated Glass Bottle
18612330 20 L Poly Drum
18612331 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612332 Supplier Spectrum Chemical Mfg Cor Supplier No. P1481500MLGL
Only null left

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity ∼75%
Grade Technical
Chemical Name or Material Phosphoric Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.