missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 20% (v/v), Ricca Chemical

$86.97 - $893.44
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H3O4P |
Molecular Weight (g/mol) | 97.994 |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforicoShow More |
PubChem CID | 1004 |
ChEBI | CHEBI:26078 |
IUPAC Name | phosphoric acid |
SMILES | OP(=O)(O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
58503-16
|
Ricca Chemical Company
5850316 |
500 mL | Natural Poly Bottle |
Each for $86.97
|
|
|||||
58503-32
|
Ricca Chemical Company
5850332 |
1 L | Natural Poly Bottle |
Each for $182.91
|
|
|||||
R58503004A
|
Ricca Chemical Company
R58503004A |
4 L | Natural Poly Bottle |
Each for $332.03
|
|
|||||
R585030020F
|
Ricca Chemical Company
R585030020F |
20 L | Cubitainer |
Each for $893.44
|
|
|||||
Chemical Identifiers
7732-18-5 | |
97.994 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforicoShow More | |
CHEBI:26078 | |
OP(=O)(O)O |
H3O4P | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
1004 | |
phosphoric acid |
Specifications
Colorless | |
79.0,19.0 | |
Natural Poly Bottle | |
H3O4P | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
Laboratory | |
Phosphoric Acid |
7732-18-5,7664-38-2 | |
81.0,21.0 | |
500 mL | |
Liquid | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
20% V/V | |
20% (v/v) Aqueous Solution (1 + 4) |
Safety and Handling
ShelfLife : 24 months