missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 20% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company 5850332
Specifications
| Phosphoric Acid | |
| 7732-18-5,7664-38-2 | |
| 81.0,21.0 | |
| 1 L | |
| Liquid | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid | |
| 1004 | |
| 20% V/V | |
| 20% (v/v) Aqueous Solution (1 + 4) |
| Colorless | |
| 79.0,19.0 | |
| Natural Poly Bottle | |
| H3O4P | |
| orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
| OP(=O)(O)O | |
| 97.994 | |
| CHEBI:26078 | |
| Laboratory |
Chemical Identifiers
| 7732-18-5 | |
| 97.994 | |
| orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
| CHEBI:26078 | |
| OP(=O)(O)O |
| H3O4P | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| 1004 | |
| phosphoric acid |
Safety and Handling
ShelfLife : 24 months