missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 10% (v/v), Ricca Chemical
$102.50 - $403.63
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | HO3P |
| Molecular Weight (g/mol) | 79.979 |
| InChI Key | UEZVMMHDMIWARA-UHFFFAOYSA-N |
| Synonym | metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate |
| PubChem CID | 3084658 |
| SMILES | OP(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
5850-16
|
Ricca Chemical Company
585016 |
500 mL | Natural Poly Bottle |
Each for $102.50
|
|
||||
|
5850-32
|
Ricca Chemical Company
585032 |
1 L | Natural Poly Bottle |
Each for $163.77
|
|
||||
|
5850-1
|
Ricca Chemical Company
RI58501 |
4 L | Natural Poly Bottle |
Each for $403.63
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| 79.979 | |
| metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
| OP(=O)=O |
| HO3P | |
| UEZVMMHDMIWARA-UHFFFAOYSA-N | |
| 3084658 |
Specifications
| 7732-18-5,7664-38-2 | |
| Aqueous Solution | |
| HO3P | |
| UEZVMMHDMIWARA-UHFFFAOYSA-N | |
| 79.979 | |
| 10% V/V | |
| 10% (v/v) |
| Natural Poly Bottle | |
| 500 mL | |
| metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
| OP(=O)=O | |
| 3084658 | |
| Laboratory | |
| Phosphoric Acid |