missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 10% (v/v), Ricca Chemical

Catalog No. 585016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
585016 500 mL Natural Poly Bottle
585032 1 L Natural Poly Bottle
58501 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 585016 Supplier Ricca Chemical Company Supplier No. 585016

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Percent Purity 10% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (v/v)
Chemical Name or Material Phosphoric Acid