missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Standard, 500 ppm PO43- (163 ppm P), Ricca Chemical

Catalog No. 583816
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
583816 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 583816 Supplier Ricca Chemical Company Supplier No. 583816
Only null left

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Amber Glass Bottle
Equivalence 1mL = 0.5mg PO4,500ppm PO4,163ppm Phosphorus (P)
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Phosphate Standard