missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade, Spectrum™ Chemical

Catalog No. 18605162
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18605162 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18605162 Supplier Spectrum Chemical Mfg Cor Supplier No. P3033500ML
Only null left

P3033

Spectrum™ Chemical Phosphate Buffered Saline Solution, 1X, Sterile, BiotechGrade is abbreviated as PBS and is used in biological research as a buffer solution containing sodium chloride and sodium phosphate, as well as potassium phosphate and potassium chloride in some formulations. It is also used in cell container rinsing and substance dilution. Spectrum offers highly pure reagents suitable for biochemical research and analysis.

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 1X
pH 7.3 to 7.5
Quantity 500 mL
Packaging Poly Bottle
Sterility Sterile
Grade Biotech