Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), with Triton∣r X-100
Ideal for maintaining a constant pH.
Supplier:  Thermo Scientific Chemicals J63521AK
 View more versions of this product
Catalog No. AAJ63521AK
Only null left
Explore available promotions
Description
Description
Phosphate-buffered saline is used in biological research. PBS buffer is ideal for maintaining a constant pH, and since it is isotonic to cells. PBS is frequently used as a wash and dilution buffer, and is also used to take a reference spectrum when measuring protein adsorption. And also used in immunoassay procedures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Phosphate Buffered Saline (PBS)
10 X
Liquid
250 mL
Product Suggestions
Product Suggestions