Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), RNase free
Helps to maintain a constant pH
Supplier:  Thermo Scientific Chemicals J62851AP
 View more versions of this product
Catalog No. AAJ62851AP
Only null left
Explore available promotions
Description
Description
Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. Phosphate-buffered saline with EDTA is used to disengage attached and clumped cells. It is also used in cell culturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Phosphate Buffered Saline (PBS)
10 X
Liquid
500 mL
Product Suggestions
Product Suggestions