missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), pH 7.6

Catalog No. AAJ62692K7
Change view
Click to view available options
Quantity:
1 L
2 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62692K7 4 L
AAJ62692K2 1 L
AAJ62692K3 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62692K7 Supplier Thermo Scientific Chemicals Supplier No. J62692K7
Only null left

Helps to maintain a constant pH

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
pH 7.6
Physical Form Liquid
Quantity 4 L

RUO – Research Use Only