Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula
Catalog No. AAJ61917K3
Change view
Click to view available options
Quantity:
2 L
Catalog No. Quantity
AAJ61917K3 2 L
1 options
Catalog No. AAJ61917K3 Supplier Thermo Scientific Chemicals Supplier No. J61917K3
Only null left
Used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells.
Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula is used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells. It plays an important role in the transportation of tissue samples and as diluents in various reagent preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form Liquid
Quantity 2 L
Packaging Plastic bottle
RUO – Research Use Only