missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Phosphate Buffered Saline
Description
Phosphate Buffered Salts, Dulbecco's Formula (DPBS) is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells. DPBS is used to irrigate, wash, and dilute mammalian cells. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Quantity | 10 L |
| Solution Type | Phosphate Buffered Saline |
| Grade | Not Available |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.