missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffered Saline, 0.01 M, pH 7.40 at 25°C, Ricca Chemical

Catalog No. R582350010F
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R582350010F 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R582350010F Supplier Ricca Chemical Company Supplier No. R582350010F
Only null left

Phosphate Buffered Saline, 0.01 M, pH 7.40 at 25°C is non-Sterile.

  • Non-sterile saline

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Melting Point ∼ 0°C
Color Colorless
Concentration 0.01 M
For Use With (Application) in Vitro Diagnostics
pH 7.4
Physical Form Solution
Quantity 10 L
Solution Type Phosphate Buffered Saline
Molecular Formula H2KO4P
Packaging Cubitainer
MDL Number MFCD00011401 MFCD00147253
Sterility Non-sterile
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
Grade Chemical Solution
Show More Show Less