missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate Buffer pH 4.1, For Cyanide, Certified, LabChem™
Supplier: LabChem LC185502
Specifications
| Phosphate Buffer pH 4.1 | |
| Colorless | |
| 4.1 ±0.3 | |
| 1 L | |
| NaH2PO4 | |
| Traceable to NIST | |
| 119.98 |
| For Cyanide | |
| Phosphine; Phosphorus oxides; Phosgene | |
| Liquid | |
| Passes Test | |
| Poly Bottle | |
| Soluble in water | |
| Certified |