missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer pH 4.1, For Cyanide, Certified, LabChem™

Catalog No. p-5388058
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC185501 500 mL
LC185502 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC185501 Supplier LabChem Supplier No. LC185501
Only null left

Specifications

Chemical Name or Material Phosphate Buffer pH 4.1
Name Note For Cyanide
CAS 7558-80-7, 7732-18-5
Color Colorless
CAS Max % 88, 12
Decomposition Information Phosphine; Phosphorus oxides; Phosgene
pH 4.1 ±0.3
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula NaH2PO4
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 119.98
Grade Certified
Show More Show Less