missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer, 0.025 M, pH 2.5, Ricca Chemical

Catalog No. R57870001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R57870001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R57870001A Supplier Ricca Chemical Company Supplier No. R57870001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate buffer solution for general laboratory use.

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Color Clear colorless liquid
Concentration 0.025 M
For Use With (Application) Water Analysis
pH 2.5
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Odor Odorless
Packaging Natural Poly Bottle
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Show More Show Less
Recommended Storage General