missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Phorbol 12,13-dibutyrate

Catalog No. 361063847
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361063847 10 mg
41531 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361063847 Supplier Tocris Bioscience™ Supplier No. 4153/10

Protein kinase C activator

Phorbol 12,13-dibutyrate is a protein kinase C activator. Induces contraction of vascular smooth muscle and inhibits MLC phosphatase (MLCP) in vascular smooth muscle. Activity does not alter intracellular Ca2+ concentration. Also inhibits the activity of Na+,K+ ATPase in OK cells.

For more information about how Phorbol 12,13-dibutyrate may be used, see our protocol:

Generation of β cells from hPSCs

Chemical Identifiers

CAS 37558-16-0
Molecular Formula C28H40O8

Specifications

Quantity 10 mg
Formula Weight 504.61
Purity >98%
Molecular Weight (g/mol) 504.62
Chemical Name or Material Phorbol 12, 13-dibutyrate
Recommended Storage Store at -20°C