missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Phorbol 12,13-dibutyrate
Description
Phorbol 12,13-dibutyrate is a protein kinase C activator. Induces contraction of vascular smooth muscle and inhibits MLC phosphatase (MLCP) in vascular smooth muscle. Activity does not alter intracellular Ca2+ concentration. Also inhibits the activity of Na+,K+ ATPase in OK cells.
For more information about how Phorbol 12,13-dibutyrate may be used, see our protocol:
Generation of β cells from hPSCsSpecifications
Specifications
| Quantity | 1 mg |
| Product Line | Tocris |
| Formula Weight | 504.61 |
| Purity | >98% |
| Molecular Weight (g/mol) | 504.62 |
| Chemical Name or Material | Phorbol 12, 13-dibutyrate |
Safety and Handling
| Recommended Storage | Store at -20°C |