missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Hot Start II PCR Master Mixes

Catalog No. F125S
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Color:
Colorless
Green
No. of Reactions:
1000 Reactions
200 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Reactions
F125S Colorless 200 Reactions
F125L Colorless 1000 Reactions
F126L Green 1000 Reactions
F126S Green 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Catalog No. F125S missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' F125S

Thermo Scientific Phire Hot Start II PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2.

Thermo Scientific Phire Hot Start II PCR Master Mix is a convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides, and optimized reaction buffer, including MgCl2. Only template and primers need to be added to the PCR reaction.

Phire Green Hot Start II PCR Master Mix also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Phire Hot Start II DNA Polymerase, included in the master mix, is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain that allows short extension times (10–15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).

Features

  • No reactivation step with quick hot start
  • Amplify four times faster than with hot start Taq
  • Minimal reaction optimization due to high inhibitor tolerance
  • Abundant products due to high efficiency
  • Amplify significantly longer DNA fragments than with any hot start Taq
  • Direct loading on gel with Green format

Applications

  • Hot Start PCR
  • Routine PCR
  • Non-high-fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping

Notes

  • The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.

Specifications

Concentration 2X
Content And Storage • 2X Phire Hot Start II PCR Master Mix, 4 x 1.25 mL
• DMSO, 1 x 0.5 mL
• Nuclease-free water, 4 x 1.25 mL

Store at -20°C.
GC-Rich PCR Performance Medium
Polymerase Phire Hot Start II DNA Polymerase
Reaction Speed Fast
Product Type Hot Start PCR Master Mix
Quantity 200 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Cloning, Hot-start PCR
Fidelity (vs. Taq) 2X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Starting Material DNA
Color Colorless
Show More Show Less
Do Phire and Phusion Hot Start II DNA Polymerases need a separate activation step in the PCR protocol?

No separate activation step is required since Phire and Phusion Hot Start II DNA Polymerases are inactivated by a reversibly bound, specific Affibody ligand that dissociates during initial denaturation.

For Research Use Only. Not for use in diagnostic procedures.