missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Hot Start II DNA Polymerase

Thermo Scientific Phire Hot Start II DNA Polymerase is an enhanced PCR enzyme for routine and high throughput PCR applications. It is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields.

Supplier:  Thermo Scientific™ F122S

 View more versions of this product

Catalog No. F122S


Only null left

Description

Description

Thermo Scientific Phire Hot Start II DNA Polymerase is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10–15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).

The hot start modification of Phire Hot Start II DNA Polymerase is based on the Affibody inactivation technology. This technology increases the specificity of the PCR reaction with no additional time required for initial activation of the enzyme.

Phire Green format is a combination of Phire Hot Start II DNA Polymerase and 5X Phire Green Buffer. The buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Features

  • No reactivation step with hot start
  • Amplification four times faster than hot start Taq
  • Minimal reaction optimization due to high inhibitor tolerance
  • Abundant products due to high efficiency
  • Amplifies significantly longer DNA fragments than hot start Taq
  • Direct loading on gel with Green Buffer

Applications

  • Hot Start PCR
  • Standard PCR
  • Non-high-fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping

Notes

  • The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
Specifications

Specifications

• Phire Hot Start II DNA Polymerase, 1 x 200 μL
• 5X Phire Reaction Buffer, 2 x 1.5 mL
• DMSO, 1 x 500 μL

Store at -20°C.
Phire Hot Start II DNA Polymerase
DNA Polymerase
200 reactions
Cloning, Hot-start PCR
Built-In Hot Start
Blunt
DNA
Colorless
Medium
Fast
DHPLC
Wet or Dry Ice
2X
200 Reactions
Separate Components
50 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.