missing translation for 'onlineSavingsMsg'
Learn More

Phenylurethane 99.0+%, TCI America™

Catalog No. p-7134937
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P109025G 25 g
P1090500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P109025G Supplier TCI America Supplier No. P109025G
Only null left

Chemical Identifiers

CAS 101-99-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00026806
InChI Key LBKPGNUOUPTQKA-UHFFFAOYSA-N
Synonym n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
PubChem CID 7591
IUPAC Name ethyl N-phenylcarbamate
SMILES CCOC(=O)NC1=CC=CC=C1

Specifications

Melting Point 52°C
Color White
Boiling Point 237°C
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylurethane
RTECSNumber FD8925000
TSCA Yes