accessibility menu, dialog, popup

UserName

H3COOHN

CAS RN 101-99-5

IUPAC Name: ethyl N-phenylcarbamate
Synonyms: N-phenyl ethylcarbamate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: LBKPGNUOUPTQKA-UHFFFAOYSA-N
SMILES: CCOC(=O)NC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)