missing translation for 'onlineSavingsMsg'
Learn More

(Phenylthio)acetonitrile, 98%

Catalog No. AAA1286922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1286922 100 g
AAA1286914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1286922 Supplier Thermo Scientific Chemicals Supplier No. A1286922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5219-61-4 | C8H7NS | 149.21 g/mol

Applications include, electron-deficient aromatics undergo vicarious nucleophilic substitution (VNS) reactions with the anion. Thus the molecule is equivalent to an acetonitrile anion synthon with thiophenate as the leaving group. For example, in the presence of solid NaOH in DMSO, nitrobenzene gives predominantly 4-nitrophenylacetonitrile, along with some of the 2-isomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applications include, electron-deficient aromatics undergo vicarious nucleophilic substitution (VNS) reactions with the anion. Thus the molecule is equivalent to an acetonitrile anion synthon with thiophenate as the leaving group. For example, in the presence of solid NaOH in DMSO, nitrobenzene gives predominantly 4-nitrophenylacetonitrile, along with some of the 2-isomer.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5219-61-4
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
MDL Number MFCD00019831
InChI Key QMJWPWCKRKHUNY-UHFFFAOYSA-N
Synonym phenylthio acetonitrile, phenylthioacetonitrile, 2-phenylthio acetonitrile, acetonitrile, phenylthio, phenylmercaptoacetonitrile, cyanomethyl phenyl sulfide, 2-phenylsulfanyl acetonitrile, 2-phenylthioethanenitrile, acmc-209ky7, phenylmercapto acetonitrile
PubChem CID 78891
IUPAC Name 2-phenylsulfanylacetonitrile
SMILES N#CCSC1=CC=CC=C1

Specifications

Density 1.142
Boiling Point 146°C to 147°C (14 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.582
Quantity 100 g
Beilstein 2042078
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material (Phenylthio)acetonitrile
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 226-013-3
RTECSNumber AM1900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only