accessibility menu, dialog, popup

UserName

CAS RN 5219-61-4

IUPAC Name: 2-(phenylsulfanyl)acetonitrile
Synonyms: acetonitrile, (phenylthio)-
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: QMJWPWCKRKHUNY-UHFFFAOYSA-N
SMILES: N#CCSC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)