missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Phenylthio)acetic Acid 98.0+%, TCI America™
$154.02 - $154.02
Chemical Identifiers
| CAS | 103-04-8 |
|---|---|
| Molecular Formula | C8H7O2S |
| Molecular Weight (g/mol) | 167.20 |
| MDL Number | MFCD00004355 |
| InChI Key | MOTOSAGBNXXRRE-UHFFFAOYSA-M |
| Synonym | phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide |
| PubChem CID | 59541 |
| IUPAC Name | 2-(phenylsulfanyl)acetate |
| SMILES | [O-]C(=O)CSC1=CC=CC=C1 |
Chemical Identifiers
| 103-04-8 | |
| 167.20 | |
| MOTOSAGBNXXRRE-UHFFFAOYSA-M | |
| 59541 | |
| [O-]C(=O)CSC1=CC=CC=C1 |
| C8H7O2S | |
| MFCD00004355 | |
| phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide | |
| 2-(phenylsulfanyl)acetate |
Specifications
| 103-04-8 | |
| White-Yellow | |
| MFCD00004355 | |
| phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide | |
| [O-]C(=O)CSC1=CC=CC=C1 | |
| 167.20 | |
| 168.21 | |
| Crystalline Powder |
| 64°C | |
| C8H7O2S | |
| 25 g | |
| MOTOSAGBNXXRRE-UHFFFAOYSA-M | |
| 2-(phenylsulfanyl)acetate | |
| 59541 | |
| ≥98.0% (T) | |
| (Phenylthio)acetic Acid |
Safety and Handling
EINECSNumber : (3)-1103
TSCA : No