Learn More
(Phenylthio)acetic acid, 97%
CAS: 103-04-8 | C8H7O2S | 167.20 g/mol
$106.44 - $317.75
Chemical Identifiers
| CAS | 103-04-8 |
|---|---|
| Molecular Formula | C8H7O2S |
| Molecular Weight (g/mol) | 167.20 |
| MDL Number | MFCD00004355 |
| InChI Key | MOTOSAGBNXXRRE-UHFFFAOYSA-M |
| Synonym | phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide |
| PubChem CID | 59541 |
| IUPAC Name | 2-phenylsulfanylacetic acid |
| SMILES | [O-]C(=O)CSC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2228014
|
Thermo Scientific Chemicals
B2228014 |
25 g |
N/A
|
|
|||||
|
AAB2228022
|
Thermo Scientific Chemicals
B2228022 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 103-04-8 | |
| 167.20 | |
| MOTOSAGBNXXRRE-UHFFFAOYSA-M | |
| 59541 | |
| [O-]C(=O)CSC1=CC=CC=C1 |
| C8H7O2S | |
| MFCD00004355 | |
| phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide | |
| 2-phenylsulfanylacetic acid |
Specifications
| 103-04-8 | |
| Stench | |
| MFCD00004355 | |
| 2045072 | |
| MOTOSAGBNXXRRE-UHFFFAOYSA-M | |
| 2-phenylsulfanylacetic acid | |
| 59541 | |
| 97% |
| 61°C to 66°C | |
| C8H7O2S | |
| 25 g | |
| phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide | |
| [O-]C(=O)CSC1=CC=CC=C1 | |
| 167.20 | |
| 168.22 | |
| (Phenylthio)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 203-073-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only