missing translation for 'onlineSavingsMsg'
Learn More

(Phenylthio)acetic acid, 97%

Catalog No. AAB2228014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2228014 25 g
AAB2228022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2228014 Supplier Thermo Scientific Chemicals Supplier No. B2228014

CAS: 103-04-8 | C8H7O2S | 167.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-04-8
Molecular Formula C8H7O2S
Molecular Weight (g/mol) 167.20
MDL Number MFCD00004355
InChI Key MOTOSAGBNXXRRE-UHFFFAOYSA-M
Synonym phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide
PubChem CID 59541
IUPAC Name 2-phenylsulfanylacetic acid
SMILES [O-]C(=O)CSC1=CC=CC=C1

Specifications

Melting Point 61°C to 66°C
Odor Stench
Quantity 25 g
Beilstein 2045072
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material (Phenylthio)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-073-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only