missing translation for 'onlineSavingsMsg'
Learn More

Phenylsulfonylacetonitrile 98.0+%, TCI America™

Catalog No. p07995g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07995G 5 g
P079925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07995G Supplier TCI America Supplier No. P07995G
Only null left

Chemical Identifiers

CAS 7605-28-9
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD00007550
InChI Key ZFCFFNGBCVAUDE-UHFFFAOYSA-N
Synonym benzenesulfonylacetonitrile, phenylsulfonyl acetonitrile, 2-phenylsulfonyl acetonitrile, phenylsulfonylacetonitrile, 2-benzenesulfonyl acetonitrile, phenylsulphonylacetonitrile, acetonitrile, phenylsulfonyl, benzenesulphonyl acetonitrile, benzenesulfonyl-acetonitrile, 2-phenylsulfonyl ethanenitrile
PubChem CID 82077
IUPAC Name 2-(benzenesulfonyl)acetonitrile
SMILES C1=CC=C(C=C1)S(=O)(=O)CC#N

Specifications

Melting Point 112°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 181.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylsulfonylacetonitrile