accessibility menu, dialog, popup

UserName

CAS RN 7605-28-9

IUPAC Name: 2-(benzenesulfonyl)acetonitrile
Synonyms: 2-(benzenesulfonyl)acetonitrile
Molecular Weight (g/mol): 181.21
Molecular Formula: C8H7NO2S
InChi Key: ZFCFFNGBCVAUDE-UHFFFAOYSA-N
SMILES: O=S(=O)(CC#N)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results