missing translation for 'onlineSavingsMsg'
Learn More

Phenylpyruvic Acid 93.0+%, TCI America™

Catalog No. p-7134453
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01545G 5 g
P015425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P01545G Supplier TCI America Supplier No. P01545G
Only null left

Chemical Identifiers

CAS 156-06-9
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002589
InChI Key BTNMPGBKDVTSJY-UHFFFAOYSA-N
Synonym phenylpyruvic acid, 3-phenylpyruvic acid, phenylpyruvate, keto-phenylpyruvate, beta-phenylpyruvic acid, b-phenylpyruvic acid, pyruvic acid, phenyl, 3-phenyl-2-oxopropanoate, alpha-ketohydrocinnamic acid, benzenepropanoic acid, .alpha.-oxo
PubChem CID 997
ChEBI CHEBI:30851
IUPAC Name 2-oxo-3-phenylpropanoic acid
SMILES OC(=O)C(=O)CC1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥93.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylpyruvic Acid
TSCA Yes
Recommended Storage Freezer