accessibility menu, dialog, popup

UserName

CAS RN 156-06-9

IUPAC Name: 2-oxo-3-phenylpropanoic acid
Synonyms: 3-phenylpyruvic acid keto-phenylpyruvic acid phenylpyruvic acid phenylbrenztraubensaeure
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: BTNMPGBKDVTSJY-UHFFFAOYSA-N
SMILES: OC(=O)C(=O)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results