missing translation for 'onlineSavingsMsg'
Learn More

Phenylmagnesium Chloride (27% in Tetrahydrofuran, ca. 2mol/L), TCI America™

Catalog No. P1381250G
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1381250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1381250G Supplier TCI America Supplier No. P1381250G
Only null left

Chemical Identifiers

CAS 100-59-4
Molecular Formula C6H5ClMg
Molecular Weight (g/mol) 136.86
MDL Number MFCD00000464
InChI Key GQONLASZRVFGHI-UHFFFAOYSA-M
Synonym phenylmagnesium chloride, chlorophenylmagnesium, magnesium, chlorophenyl, phenylmagnesiumchloride, chloro phenyl magnesium, phenyl magnesium chloride, phenyl-magnesium chloride, magnesium,chlorophenyl, phenylmagnesium chloride solution, 2.0 m in thf, phmgcl
PubChem CID 7513
IUPAC Name chloro(phenyl)magnesium
SMILES Cl[Mg]C1=CC=CC=C1

Specifications

UN Number 2924
Formula Weight 136.86
Quantity 250 g
Chemical Name or Material Phenylmagnesium Chloride (27% in Tetrahydrofuran, ca. 2mol/L)
EINECSNumber (3)-3903
TSCA Yes