CAS RN 100-59-4
IUPAC Name:
chloro(phenyl)magnesium
Synonyms:
phenylmagnesium chloride
Molecular Weight (g/mol):
136.86
Molecular Formula:
C6H5ClMg
InChi Key:
GQONLASZRVFGHI-UHFFFAOYSA-M
SMILES:
Cl[Mg]C1=CC=CC=C1
1
–
1
of
1
results
1
Phenylmagnesium Chloride (27% in Tetrahydrofuran, ca. 2mol/L), TCI America™
CAS: 100-59-4 Molecular Formula: C6H5ClMg Molecular Weight (g/mol): 136.86 MDL Number: MFCD00000464 InChI Key: GQONLASZRVFGHI-UHFFFAOYSA-M Synonym: phenylmagnesium chloride,chlorophenylmagnesium,magnesium, chlorophenyl,phenylmagnesiumchloride,chloro phenyl magnesium,phenyl magnesium chloride,phenyl-magnesium chloride,magnesium,chlorophenyl,phenylmagnesium chloride solution, 2.0 m in thf,phmgcl PubChem CID: 7513 IUPAC Name: chloro(phenyl)magnesium SMILES: Cl[Mg]C1=CC=CC=C1